missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD MDCK Cell Lysate
Convenient to use and useful control for western blotting application
Brand: BD 611635
Additional Details : Weight : 0.01100kg
Description
- Source is rich in endogenously expressed proteins
- Provided in denatured format allowing for immediate loading onto SDS-PAGE gel
- Storage buffer: SDS-PAGE buffer (62mM Tris pH 6.8, 2% SDS, 0.9% b-mercaptoethanol, 0.003% bromophenol blue, 5% glycerol)
- Concentration: 1mg/mL
Routinely tested western blot
Specifications
MDCK | |
1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol | |
1 mg/mL |
500 μg | |
Canine | |
-20°C |
Research use only