Learn More
BD Microtainer™ Capillary Blood Collector and BD Microgard™ Closure
Textured inner surface and large tube diameter to aid sample mixing
Brand: BD 365974
Additional Details : Weight : 0.05000kg
Description
- Designed to minimize sample contact and improve mixing
- “Twist assist” feature allows controlled closure removal; snap-lock fit assures complete seal
- Color-coded closures for accurate tube selection
- 8mm dia. for easy probe and pipet access; integral collector eliminates need for assembly
- Marked with fill lines, lot numbers and expiration date
- Holds 250 to 500μL of blood
- Extender (optional) facilitates labeling and handling
Specifications
Dipotassium EDTA Tube | |
250 to 500 μL | |
Lavender |
Microgard | |
8 mm |