missing translation for 'onlineSavingsMsg'
Learn More

BD MitoScreen Flow Cytometry Mitochondrial Membrane Potential Detection Kit

Contains JC-1, which has been optimized for use in flow cytometry, and 10 assay buffer

Brand:  BD 551302

Additional Details : Weight : 0.50000kg

Product Code. 15896598

  • £484.00 / 1 set
Estimated Shipment: 07-08-2024
to see stock.

Includes: JC-1 Dye and Assay Buffer

Description

Description

  • Contains 100% JC-1 (5,5',6,6'-tetrachloro-1,1',3,3'-tetraethylbenzimidazolcarbocyanine iodide) dye, an irritant
  • JC-1, typically excited using 488nm line of an argon ion laser while the monomeric form emits at 527nm and J-aggregates at 590nm

Specifications

Specifications

JC-1 dye and assay buffer: 100% JC-1 (5,5',6,6'-tetrachloro-1,1',3,3'-tetraethylbenzimidazolcarbocyanine iodide) dye, an irritant
2° to 8°C
RUO
Flow Cytometer
100
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers