missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Mouse Cerebrum Lysate
Convenient to use and provided in denatured format allowing for immediate loading onto SDS-PAGE gel
Brand: BD 611455
Additional Details : Weight : 0.01100kg
Includes: Mouse Cerebrum
Description
- Useful control for western blotting application
- Rich in endogenously expressed proteins
Specifications
Mouse Cerebrum | |
1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol | |
1 mg/mL |
500 μg | |
Mouse | |
-20°C |