Learn More
BD Mouse IL-4 ELISA Set
An immunoassay with different sensitivity and dynamic range
Brand:  BD 555232
Additional Details : Weight : 0.14400kg
Product Code. 15838028
  • £1205.00 / 1 set
Estimated Shipment: 04-09-2024
to see stock.
Add to basket
Includes: Capture antibody, detection antibody, enzyme reagent, standards, instruction/analysis certificate
Description
Description
To develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant mouse IL-4 in serum, plasma, and cell culture supernatants
Enzyme-linked immunosorbent assay
Specifications
Specifications
7.8 to 500pg/mL
20 96-well Plates
anti-Mouse IL-4 monoclonal (capture) antibody, Biotinylated anti-Mouse IL-4 monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Mouse IL-4 (standard), Streptavidin-HRP Conjugate
2°C to 8°C
Mouse IL-4 ELISA set
To develop ELISA for natural or recombinant Mouse IL-4 in Serum, Plasma and cell culture supernatants
20 Plates
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Research Use Only