missing translation for 'onlineSavingsMsg'
Learn More

BD Mouse intracellular CytoKine-1 Positive Control Cells, MiCK-1

Contains ∽5 x 106 fixed, non-permeabilized mouse lymphoid cells

Brand:  BD 554652

Product Code. 15887818

  • £720.00 / Each
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Cells easily express detectable levels of intracellular IL-2, TNF and IFN-Λ
  • Prepared by stimulating mouse spleen cells in the presence of a protein transport inhibitor
  • Contains a measurable proportion of cytokine-producing cells
  • Storage buffer: frozen in FBS and 10% DMSO
    • Intracellular staining (flow cytometry)

      TRUSTED_SUSTAINABILITY
Specifications

Specifications

Light Brown
Frozen
Intracellular staining
1 mL
5 x 106 cells/mL
IL-2, TNF and IFN-L
≈5 x 106 Activated Mouse splenocytes (20 test samples at 2.5 x 105 cells/test) fixed and suspended in 1mL
-80°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For research use only.