missing translation for 'onlineSavingsMsg'
Learn More

BD Mouse intracellular CytoKine-3 Positive Control Cells, MiCK-3

Contains fixed,non-permeabilized mouse lymphoid cells

Brand:  BD 554654

Additional Details : Weight : 0.01000kg

Product Code. 15807828

  • £1430.00 / Each
Estimated Shipment: 31-07-2024
to see stock.
Add to basket

Description

Description

  • Cells easily detects levels of intracellular IL-1α, IL-6, IL-12, MCP-1 or TNF
  • Contains measurable proportions of cytokine-producing cells
  • Storage buffer: frozen in FBS and 10% DMSO

Intracellular staining (flow cytometry)

Specifications

Specifications

Frozen
5 x 106 cells/mL
Intracellular staining
1 mL
Light Brown
IL-1alpha, IL-6, IL-12, MCP-1 or TNF
≈5 x 106 Activated Mouse macrophages (20 test samples at 2.5 x 105 cells/test) fixed and suspended in 1mL
-80°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For research use only.