Learn More
Perm Buffer IV (10x), BD
For improved immunofluorescent staining and detection of certain intracellular phosphorylated proteins
Brand:  BD Biosciences 560746
Additional Details : Weight : 0.10000kg
Product Code. 15830459
  • £335.00 / Each
Estimated Shipment: 29-07-2024
to see stock.
Description
Description
BD Phosflow™ Perm Buffer IV (Perm Buffer IV) is intended for the permeabilization of paraformaldehyde-fixed single-cell suspensions prepared from lymphoid tissues, peripheral blood mononuclear cells (PBMC) and lysed whole blood cells.
  • Treatment with buffer and subsequent washing with staining buffer enables access for fluorescent antibodies to stain intracellular molecules
  • Buffer preserves light scattering properties of cells
  • Providies capacity for intracellular immunofluorescent staining in preparation for Phosflow cytometric analysis
Flow Cytometry, Intracellular staining
Specifications
Specifications
Cell Permeabilization Buffer
10X
Permeabilization of paraformaldehyde-fixed single-cell suspensions prepared from lymphoid tissues, peripheral blood mononuclear cells and lysed whole blood cells, flow cytometry, intracellular staining
50 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Research Use Only