missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perm / Wash™ Buffer I (10x), BD
Click to view available options
Quantity:
125 mL
Unit Size:
Each
Description
Permeabilize cells and serve as an antibody diluent and cell wash buffer
- Storage buffer: Aqueous buffered solution containing saponin, fetal bovine serum and ≤0.09% sodium azide
Flow cytometry
Specifications
Specifications
| Chemical Name or Material | Perm/Wash Buffer I |
| Concentration | 10X |
| For Use With (Application) | Intracellular staining of post-translationally modified signaling Proteins to permeabilize cells, to serve as an antibody diluent, cell Wash Buffer, flow cytometry, used to permeablize or wash cells and to dilute antibodies for immunofluorescent |
| Quantity | 125 mL |
Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction