missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD PFSK-1 Cell Lysate
Convenient to use and provided in a denatured format allowing for immediate loading onto an SDS-PAGE gel
Brand: BD 611942
Additional Details : Weight : 0.01000kg
Includes: Rat Primitive Neuroectodermal Tumor, Brain
Description
- Useful control for western blotting application
- Can be rich in endogenously expressed proteins
Specifications
PFSK-1 | |
1mg/mL in SDS-PAGE Buffer: 62mM Tris pH 6.8, 2% SDS, 0.9% beta-mercaptoethanol, 0.003% Bromophenol Blue, 5% Glycerol | |
1 mg/mL |
500 μg | |
Rat | |
-20°C |