Learn More
BD Phosflow™ Violet Fluorescent Cell Barcoding Kit
Provides reagents for successfully barcoding T cell subsets from human whole blood samples for studying phosphorylated protein induction in MAP Kinase and JAK/STAT signaling pathways
Brand:  BD 561570
Additional Details : Weight : 0.01000kg
Product Code. 15851139
  • £2135.00 / Each
Estimated Shipment: 03-09-2024
to see stock.
Includes: Cell Barcoding Dye 450 (CBD450) and Cell Barcoding Dye 500 (CBD500), Fluorescent Cell Barcoding Wash Buffer, and choice of one Alexa Fluor™ 647-conjugated, phospho-specific antibody
Description
Description
  • Reactivity: human
  • Size: 800 tests
Specifications
Specifications
800 Tests