missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Phospholipid-binding Protein, APC Annexin V
Has high affinity for PS, and binds to cells with exposed PS
£535.00 - £827.00
Specifications
Sample Volume | 5 μL |
---|---|
Storage Buffer | Aqueous Buffered solution containing BSA and ≤0.09% Sodium Azide |
Content And Storage | 4°C |
For Use With (Application) | Flow Cytometry |
Conjugate | APC (Allophycocyanin) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15856278
|
BD
550474 |
100 Tests |
£535.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15866278
|
BD
550475 |
200 Tests |
£827.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Conjugated to APC
- Staining precedes the loss of membrane integrity which accompanies the latest stages of cell death resulting from either apoptotic or necrotic processes
Flow Cytometry
Specifications
5 μL | |
4°C | |
APC (Allophycocyanin) |
Aqueous Buffered solution containing BSA and ≤0.09% Sodium Azide | |
Flow Cytometry |