Learn More
Plus Permeabilization Buffer, BD
Specially formulated for the immunofluorescent staining of incorporated BrdU for flow cytometric analysis
Brand:  BD Biosciences 561651
Additional Details : Weight : 0.02400kg
Product Code. 15891199
  • £308.00 / Each
Estimated Shipment: 26-07-2024
to see stock.
Description
Description
  • Used as a staining enhancer and secondary permeabilization reagent
  • Should be used with fixed cell samples only
Flow Cytometry, Intracellular staining
Specifications
Specifications
Permeabilization Buffer
Immunofluorescent staining of incorporated BrdU for flow cytometric analysis, staining enhancer and secondary permeabilization reagent, intracellular staining
10 mL