Learn More
BD Reagent Set A
Use in BD OptEIA™ ELISA Sets
Brand: BD 550536
Additional Details : Weight : 2.60000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Includes: Substrate reagent A, substrate reagent B, wash concentrate 20X, stop solution, assay diluent, coating buffer
Description
Enzyme-linked immunosorbent assay
Specifications
OptEIA™ reagent set A | |
ELISA | |
2°C to 8°C |
20 Plates | |
ELISA Set | |
250mL Coating Buffer pH 6.5, 1L assay diluent, 1L Wash Concentrate 20X, 125mL Substrate reagent A and 125mL reagent B, 125mL sto p solution |
Safety and Handling
- OptEIA Rgnt Set A pH 6.5
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : orthophosphoric acid, sodium hydrogenorthophosphate, CMIT/MIT, methanol
Research Use Only