Learn More
BD Streptavidin Particles Plus - DM
For positive selection or depletion of leukocyte subpopulations using the BD IMagnet™
Brand: BD 557812
Additional Details : Weight : 0.02000kg
Description
The small size of BD IMag particles allows the positive fraction to be further evaluated in downstream applications.
- Magnetic nanoparticles
- Have streptavidin covalently conjugated to their surface
- Allow positive fraction to be further evaluated in downstream applications such as flow cytometry
- Storage Buffer: Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Cell separation
Specifications
5 mL | |
4°C | |
Cell separation, for positive selection or depletion of leukocyte subpopulations using the BD IMagnet |
Research use only