missing translation for 'onlineSavingsMsg'
Learn More

BD TMB Substrate Reagent Set

For use in BD OptEIA™ ELISA sets. Substrate Reagent A, which is clear and colorless, contains hydrogen peroxide. Substrate Reagent B is colorless to a very light amber solution containing 3,3', 5,5' tetramethylbenzidine (TMB) in an organic solvent.

Brand:  BD 555214

154.08 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.



Product Code. 15808028

  • £193.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes:
  • Substrate reagent A, 300mL
  • Substrate reagent B, 300mL
This item is not returnable. View return policy

Description

Description

  • Bring reagents to room temperature prior to use (allow 30 minutes for this process)
  • Discard any remaining working solutions after use
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ELISA Substrate
ELISA
2°C to 8°C
ELISA Set
Substrate reagent A, 300mL
Substrate reagent B, 300mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BD TMB Substrate Reagent Set > TMB Substrate Reagent Set

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Research Use Only