missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Eccentric Tip Syringe

Product Code. 12044717
Click to view available options
Unit Size:
Pack of 120
This item is not returnable. View return policy

Product Code. 12044717

Brand: BD 300613

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Used for medical applications. BD Medical™ Vacutainer™ Eccentric Tip Syringe is graduated with a 20mL capacity.

  • 20 mL
  • Eccentric tip
  • Graduation 1 mL
TRUSTED_SUSTAINABILITY

Specifications

Product Type Syringe
Volume (Metric) 20 mL
Connection Type Eccentric
Sterility Sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.