Learn More
Buerkle™ BeefSteaker Sampler
Buerkle™ BeefSteaker Sampler designed to take bacteriological samples from the surfaces of dead animals.
Brand:  Buerkle™ 5388-1000
230.53 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Product Code. 11816403
  • £259.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For taking bacteriological samples from the surfaces of dead animals in slaughterhouses and in butchers' shops (cattle, pigs, sheep, goats and horses)
  • Used to cut out a 25mm round sample from the animal‘s skin
  • Stainless steel AISI 420 cutter allows the sample to be cut out easily
  • Can also be used for other sampling purposes (histology, medicine, food and semi-luxury articles, cheese factories, agriculture)
  • Can be steam-sterilised or flamed
  • A hole in the crown prevents air compression and is used for ejecting the sample
Specifications
Specifications
Sampler for Meat
Autoclavable
25 mm
Stainless Steel (V2A)
Foodstuffs