Learn More
Bel-Art™ Thick Biohazard Disposal Bags, 1.5 mil
Include black biohazard symbol and precautionary procedures printed in four languages. Bags are autoclavable and are effective for the disposal of biohazardous materials.
£265.00 - £870.00
Specifications
Product Type | Biohazard Disposal Bag |
---|---|
Autoclavable | Autoclavable |
Certifications/Compliance | ASTM 1922 Tear Resistance, ASTM 1709 Dart Impact |
Closure Type | Zip Tie |
Color | Red |
Product Code | Brand | Capacity (English) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (English) | Price | Quantity & Availability | ||||||
11392154
|
Bel-Art™
F13164-1419 |
2 to 4 gal. |
£265.00
Pack of 200 |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
11302164
|
Bel-Art™
F13164-1923 |
5 to 9 gal. |
£358.00
Pack of 200 |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
11312164
|
Bel-Art™
F13164-2535 |
13 to 20 gal. |
£870.00
Pack of 200 |
Estimated Shipment: 07-07-2025
Log in to see stock. |
||||||
11737927
|
Bel-Art™
F13164-3138 |
20 to 30 gal. |
£596.00
Pack of 200 |
In Stock
Log in to see stock. |
||||||
10296674
|
Bel-Art™
F13164-3848 |
40 to 55 gal. |
£490.00
Pack of 100 |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
Description
Include black biohazard symbol and precautionary procedures printed in four languages. Bags are autoclavable and are effective for the disposal of biohazardous materials.
- Bright red polypropylene bags include black biohazard symbol
- Precautionary procedures printed on bag in English, Spanish, French, and German for added safety
- Sterilization patch darkens when subjected to steam sterilization
- High temperature steam autoclavable to 135°C (275°F)
- Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests
- Made in USA
Specifications
Biohazard Disposal Bag | |
ASTM 1922 Tear Resistance, ASTM 1709 Dart Impact | |
Red | |
Used for biohazard waste | |
Polypropylene | |
0.279 mm |
Autoclavable | |
Zip Tie | |
Single-use | |
Pre-printed sterilization indicator patch, labeled with the standard biohazard warning symbol or precautionary protocols English, French, German, Spanish | |
Non-sterile | |
1.50 mil |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.