missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Covers for 96 Deep-Well Plate

Product Code. 10350981 Shop All SP Scienceware Products
Click to view available options
Color:
Natural
Unit Size:
Pack of 24
This item is not returnable. View return policy

Product Code. 10350981

Brand: Bel-Art™ F378600004

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Covers for use with Bel-Art™ 96 Deep-Well Plates. Reduce evaporation and permit safe and stable stacking of plates.

  • Covers to be used with Bel-Art™ 96 Deep-Well Plates (F378600001, F378600000)
  • Specially designed polypropylene (PP), keyed fit cover separates each well with a sealing ring to reduce evaporation and permits safe and stable stacking of plates
  • Covers are keyed to fit only one way

Specifications

Barcode No Barcode
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Color Natural
Product Type Well Plate Lid
For Use With (Equipment) SP Scienceware 96 Deep-Well Plates (F378600001, F378600000)
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape No Bottom
For Use With (Application) Used for variety of processes and operations such as sequencing, precipitations and dilution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.