missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Accessories for Write-on Tape Dispenser
The U-shaped stainless steel adapter presents the dispenser at a 45° angle off the wall to facilitate writing
Brand: Bel-Art™ F13473-0001
This item is not returnable.
View return policy
Description
- Economy Model Dispenser (Mfr. No. F13473-0000) can be wall mounted vertically or horizontally with the platform positioned to the left or right
- Compatible with Wet Surface Pen (sold separately), which offers consistent ink flow on vertical surfaces
Specifications
Wall Mount Foot | |
Stainless Steel | |
Mounts vertically or horizontally |
Rolls of Tape | |
4 Rolls x 25 mm W (1 in.) or 5 Rolls x 12 to 19 mm W (1/2 to 3/4 in.) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction