missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Acid/Solvent Bottle Carriers
Polyethylene carriers with epoxy coated, heavy-gauge steel wire bails have wide ribs in both the sides and on the bottom to support and cushion bottles
Brand: Bel-Art™ F16957-0000
This item is not returnable.
View return policy
Description
- Offer shatter resistant protection for glass bottles
- Cone style lid has a 5/8 in. (16mm) opening in top for insertion of tubing; simplifies the transfer of liquids without pouring
- Truncated style lid permits pouring without the removal of the lid
- Designed to hold standard US reagent bottles
Specifications
Bottle Carrier | |
Bottles (2.5 L) | |
35 cm |
Not Autoclavable | |
Polypropylene | |
13.5 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction