missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Acid/Solvent Bottle Carriers

Polyethylene carriers with epoxy coated, heavy-gauge steel wire bails have wide ribs in both the sides and on the bottom to support and cushion bottles

Brand:  Bel-Art™ F16960-0000

 View more versions of this product

Product Code. 11753207

  • £119.00 / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Offer shatter resistant protection for glass bottles
  • Cone style lid has a 5/8 in. (16mm) opening in top for insertion of tubing; simplifies the transfer of liquids without pouring
  • Truncated style lid permits pouring without the removal of the lid
  • Designed to hold standard US reagent bottles

Specifications

Specifications

Bottle Carrier
Bottles (4 L)
37 cm
Not Autoclavable
Polypropylene
14.37 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers