missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Bacterial Cell Spreaders
Spreads bacterial cells onto the surface of an agar Bel Art™ SP Scienceware™ Bacterial Cell Spreaders are available in two sizes.
£32.30 - £41.35
Specifications
Autoclavable | Yes |
---|---|
Material | Stainless Steel |
Product Type | Bacterial Cell Spreader |
Sterility | Sterile |
Length (English) | 12 in. |
Product Code | Brand | For Use With (Equipment) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Length (Metric) | Price | Quantity & Availability | ||||
11326954
|
Bel-Art™
F37736-0006 |
Inoculating turntable, 6cm inoculating plates | 30.2 cm |
£32.30
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
11336954
|
Bel-Art™
F37736-0009 |
Inoculating turntable, 9cm inoculating plates | 30.2 cm |
£41.35
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
||||
Description
Spreads bacterial cells onto the surface of an agar Bel Art™ SP Scienceware™ Bacterial Cell Spreaders are available in two sizes.
- Autoclavable and may be flame sterilized
- Features triangular end with smooth rounded surfaces throughout
- Smaller 16mm width bar suitable for use with 6cm plates
- Larger 25mm width bar suitable for use with 9cm plates
- Use with Inoculating Turntable (Mfr. No.F40913-0000)
- Both sizes 30cm long
Specifications
Yes | |
Bacterial Cell Spreader | |
12 in. | |
Easily perform inoculation of petri dishes |
Stainless Steel | |
Sterile | |
30.2 cm |