Learn More
Bel-Art™ SP Scienceware™ Glove Box Accessory, Iris Port Entry
SP Scienceware™ Bare Hand Entry (Iris) Port allows bare hnd entry and operation.
Brand:  Bel-Art™ H50025-0020
Product Code. 10646103
  • £1050.00 / Pack of 2
Explore more special offers
Description
Description
  • Fits 7 3/4 in. I.D. (19.7cm) glove ports
  • Made of ten layers of soft, conforming rubber
  • Easy access and withdrawal, plus excellent sealing
Order Info
One pair per pack.
Specifications
Specifications
Silicone Latex Rubber
19.7cm I.D. glove ports
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products