missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Battery-Powered Engraving Tool and Stand

Permanently mark important tools, samples and more in seconds

Brand:  Bel-Art™ F44150-0005

 View more versions of this product

Product Code. 11376354

  • £47.25 / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Includes: Extra engraving tip

Requires : AA batteries (not included)

Description

Description

Engraving Tool (Mfr. No. F441500005)

  • Hold the Engraving Tool like a pen, press the power button, apply light pressure, and “write” to permanently mark most materials including glass, plastic, wood, metal, bone, and stone
  • Portable, cordless battery power: operates on two “AA” batteries (not included)
  • Use indoors or out in the field
  • Marks will not fade if exposed to chemicals or sunlight

Engraving Tool Stand (Mfr. No. F441500015)
  • Keeps tool handy when not in use
  • Molded from polyethylene
  • Well to hold extra engraving tip

Order Info

7 in. (17.8cm) long

Specifications

Specifications

Battery-powered Engraving Tool
7 in.
Extra engraving tip
Engrave test tubes, bottles, racks, keys, equipment, tools, electronics and more
18 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers