missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Battery-Powered Engraving Tool and Stand

Permanently mark important tools, samples and more in seconds

Brand:  Bel-Art™ F44150-0015

 View more versions of this product

Product Code. 11386354

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Includes: Extra engraving tip

Requires : AA batteries (not included)
This item is not returnable. View return policy

Description

Description

Engraving Tool (Mfr. No. F441500005)

  • Hold the Engraving Tool like a pen, press the power button, apply light pressure, and “write” to permanently mark most materials including glass, plastic, wood, metal, bone, and stone
  • Portable, cordless battery power: operates on two “AA” batteries (not included)
  • Use indoors or out in the field
  • Marks will not fade if exposed to chemicals or sunlight

Engraving Tool Stand (Mfr. No. F441500015)
  • Keeps tool handy when not in use
  • Molded from polyethylene
  • Well to hold extra engraving tip

Order Info

7 in. (17.8cm) long

Specifications

Specifications

Battery-powered Engraving Tool Stand
Polypropylene
Extra engraving tip
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers