missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Bel-Blotter™ 96-Well Replicating Tool

Designed to do tedious tasks with a minimum of effort

Brand:  Bel-Art™ F37876-0002

25.47 GBP valid until 2024-11-30
Use promo code "21681" to get your promotional price.


Product Code. 10748661

  • £32.05 / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The 96 open-ended pipette tips of this tool fit all types of 96 well plates from flat, vee or round bottom plates to 0.2mL thin walled PCR plates and tubes
  • Can be used for replication of recombinant DNA libraries, inoculation of filters for colony hybridization, PCR, phage typing and other applications
  • Made of polycarbonate plastic, it is easy-to-use, reusable and autoclavable

Specifications

Specifications

Autoclavable
Increase Productivity, Minimize Effort
0.2 mL
Polycarbonate
1 Pack
96-Well Replicating Tool
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers