missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Benchtop Biohazard Disposal Bag

Product Code. 11735084 Shop All SP Scienceware Products
Click to view available options
Dimensions (L x W):
8.5 x 12 in.
Unit Size:
Pack of 1000

Product Code. 11735084

Brand: Bel-Art™ F131660001

Only null left
Add to basket
Add to basket

For disposal of tips and tubes.

  • Bright red
  • Features one long flap for easy opening
  • Printed with biohazard symbol and sterilization indicator patch
  • Made from high molecular weight high-density polyethylene
  • Size allows bag to be inverted over hand to safely pick up waste.
  • Steam autoclavable at 121°C (250°F)

Specifications

Autoclavable Autoclavable
Capacity (English) 0.5 gal.
Capacity (Metric) 2 L
Closure Type Zip Tie
Color Red
Disposable Single-use
For Use With (Application) For disposal of tips and tubes.
Includes Bags Only
Material HDPE (High-molecular Weight)
Sterility Non-sterile
Thickness (Metric) 0.018 mm
Thickness (English) 0.72 mil
Dimensions (L x W) 8.5 x 12 in.
Length (English) 8.5 in.
Length (Metric) 21.6 cm
Width (English) 8.5 in.
Width (Metric) 22 cm
Product Type Benchtop Biohazard Disposal Bag
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.