missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Bent Handle Scoop

Features an ergonomic, curved handle design to elevate the hand from the sample, reducing risk of contamination

Brand:  Bel-Art™ H36902-2006

Code : 04

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 13452489

  • £41.10 / Pack of 10
Estimated Shipment: 03-09-2024
to see stock.
Add to basket

Description

Description

  • Curved handle helps prevent incidental contact with sample source material and exposure to hazardous substances
  • Flat bottom keeps scoop upright when placed on a flat surface; can be used as a weigh boat
  • Made from high-impact, lightweight, FDA grade polystyrene
  • Sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Sterilization
  • Individually packaged and ready for one time use and disposal

Specifications

Specifications

Scoop
Flat
2 oz.
Yes
FDA
7.62 x 1.62 x 1.12cm (19.4 x 4.1 x 2.9 cm)
1.12 in.
2.9 cm
3.06 in.
19.4 cm
11.6 cm
1.62 in.
4.1 cm
Polystyrene
Bent and curved
60 mL
Sterile
Design to elevate the hand from the sample, reducing risk of contamination, prevent incidental contact with sample source material and exposure to hazardous substances
1.12 in.
2.9 cm
7.62 in.
4.56 in.
7.62 cm
1.62 in.
4.1 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers