missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Clavies™ Biohazard Bag Holder Cover
Opens quickly and easily by pressing the built-in release tab — no batteries required
Brand: Bel-Art™ F13192-0102
36.43 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Code : 03
Additional Details : Weight : 1.40000kg
Description
- Cover installs in seconds to improve sanitation of biohazard waste containers
- Dishwasher and autoclave safe: autoclavable at 121°C (250°F)
- Fits both the medium and large sizes of the Clavies Biohazard Bag Holders (Mfr. Nos. 13192-0002 and 13192-0003)
- Features a smooth 4 x 4 in. (10.1 x 10.1cm) area for applying labels
- Center panel of the lid opens to an upright position and reveals a large 8 x 9.5 in. (20.3 x 24.1cm) opening
- Orange
Specifications
Orange | |
Improves the sanitation of biohazardous waste containers | |
Biohazard Bag Holder Cover |
Autoclavable | |
Clavies™ biohazard bag holders |