missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Cleanware™ Self-Adhesive Waste Bags

Disposable waste bags free bench or hood space when hung from walls. Bel Art™ SP Scienceware™ Cleanware™ Self-Adhesive Waste Bags adhere to cabinets and other surfaces.

£63.20 - £94.80

Specifications

Product Type Laboratory Waste Bag
Color White
Disposable Single-use
For Use With (Application) Cleanware
Includes Self-adhesive strip
View More Specs

Products 2
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
11372144
View Documents Promotion Details
Bel-Art™
F13174-1008
10 x 8 in. (25.4 x 20.32cm)
£63.20
Pack of 50
In Stock
to see stock.
Edge
Add to basket
 
11382144
View Documents Promotion Details
Bel-Art™
F13174-1012
16 x 12 in. (40.64 x 30.48cm)
£94.80
Pack of 50
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Peel backing off the adhesive strip across the top and hang close to your work area
  • Back panel is longer than the front for easy opening
  • High Molecular Weight High Density (HMHD) polyethylene; steam autoclavable at 121°C (250°F)
  • Maximum weight of contents should not exceed 1.36kg (3lb)

Compliance

Pass ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance Tests

Specifications

Specifications

Laboratory Waste Bag
Single-use
Self-adhesive strip
1 mil
White
Cleanware
HDPE
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.