missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Equilibrium Dialysis Cells
Feature stainless steel screws that tighten down on the dialysis membrane also acting as a gasket
£216.00 - £262.00
Specifications
Product Type | Dialysis Cell |
---|---|
Material | Acrylic |
Color | Transparent |
Quantity | Each |
For Use With (Application) | Stainless steel screws that tighten down on the dialysis membrane also acting as a gasket |
Description
- Made from clear acrylic
- Filler hole screws on each half cavity are fluoropolymer-coated stainless steel
- Volume shown is for one half cell (double for total cell volume)
- A red dot on one half of the equilibrium cell easily identifies the otherwise identical halves
Specifications
Dialysis Cell | |
Transparent | |
Stainless steel screws that tighten down on the dialysis membrane also acting as a gasket |
Acrylic | |
Each |