Learn More
Bel-Art™ Fire Blanket
Extinguishes small class A, light oil and grease fires without the mess of foam or chemical agents. Size: 36 x 40 in.
Brand:  Bel-Art™ H24869-0000
Product Code. 10545804
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • For small Class A oil/grease fires and flare-ups
  • May also be wrapped around the head and body for protection against flames and embers when fleeing a burning room, or for protection from splashing molten metal
  • Woven fiberglass with a textured surface; all edges are serged with fireproof Kevlar™ thread
  • Close weave greatly reduces the amount of atmospheric oxygen available to feed the fire
  • Ppackaged in an instant access container that can be hung on the wall or stored in any accessible location
Specifications
Specifications
Fire Blanket
Woven Fiberglass
Ideal for class A light Oil and grease flare-ups
91.4 cm
40 in.
Tan
36 x 40 in. (91.4 x 101.6 cm)
36 in.
0/1 V
101.6 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bel-Art™ Fire Blanket > Fire Blanket, 36 x 40 in.