missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Wash Bottles: Deionized Water
38.51 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
Capacity (Metric):
500 mL
Description
- Wide mouth of 53mm (2 in.) for easy refilling
- Pre-printed with Chemical name and formula, National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
- Translucent LDPE bottles
- Color-coded polypropylene closure
Specifications
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Includes | Pack of 4 each of “DI Water” wash botttles |
Material | LDPE |
Quantity | 4/Pack |
Closure Color | Blue |
Product Type | Wash Bottle |
Chemical Label | Deionized Water |
Closure Material | Polypropylene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'