missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Frigimat™ Dry Ice Maker

Produces dry ice in four minutes.

Brand:  Bel-Art™ F38874-0000

Product Code. 15167584

  • £621.00 / Each
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Easy and safe to use with quiet operation
  • Multiple pressure reliefs prevent pressure from exceeding 10psi
  • Built-in pressure gauge monitors dry ice production
  • Features a clear, polycarbonate body
  • Body temperature is always safe to handle without fear of freezing injury
  • Operates with CO2 cylinder (sold separately)
Specifications

Specifications

Easy and fast dry ice production on demand
10 to 16 blocks of dry ice
Polycarbonate
Dry Ice Maker
50 lb. LCO2 cylinder
1.8m Nickel plated Copper connecting tube with standard CGA 320 thread coupling for easy attachment to LCO2 cylinder, rugged Nickel plated pigtail, pressure relief safeguards
Dry Ice Maker
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Frigimat™ Dry Ice Maker > Dry Ice Maker

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.