Learn More
Bel-Art™ SP Scienceware™ Glascribe™ Pen
Permanently marks laboratory glassware, ceramics and plastics
Brand: Bel-Art™ H44150-0000
38.36 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Code : 03
Additional Details : Weight : 0.13500kg
Description
- Tungsten carbide tip (Mohs Hardness = 9) easily inscribes identifying marks on labware
- Retractible tip operates with pen style push button
- Clips to a pocket, ready for use
Specifications
Scribe Pen | |
Tungsten Carbide | |
Permanently marks laboratory glassware, ceramics and plastics |
Retractable | |
Pocket clip | |
Each |