Learn More
Bel-Art™ SP Scienceware™ Glove Box Economy Sleeve Gloves
Click to view available options
Size:
9
10
Description
- Single piece glove with sleeve
- For glove boxes with 8 in. glove ports
- Requires clamping rings (19-821-983) to hold gloves in place
- Sold in pairs
Specifications
Specifications
Size | 9 |