missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Hand-Held Colony Counter
Bel-Art™ SP Scienceware™ Hand-Held Colony Counter is simple-to-operate colony counter marks, counts and gives an audible and visual alert each time count is registered.
Brand: Bel-Art™ F37862-0002
Includes: Long-life red and black felt-tip markers, 3V lithium battery, protective storage case.
Description
- Fast, accurate and economical sample counting and sample count verification
- Stores up to 35 sample counts
- Verifies sample counts with up/down counting
- Displays sum total of all colonies counted
- Backlit display for easy viewing in low light conditions
- Audible alert can be turned off if desired
- Counting function can be locked to prevent error during breaks between counting
- Included felt-tip markers work on plastic or glass and are oil- and water-resistant; ink is removable with ethyl alcohol
Order Info
Colony Counter System (07-910-022) also available. Replacement markers (07-910-019 and 07-910-020) and battery (07-910-021) sold separately.
Specifications
Colony Counter | |
Stores up to 35 samples | |
CE | |
Long life red and black felt tip markers, 3V Lithium battery, protective storage case |
Backlit LCD | |
6 in. | |
Count colonies of Bacteria or other microorganisms grown on an agar plate |