Learn More
Bel-Art™ Lab-Aire™ II Glassware Drying Racks
Glassware and plasticware drying racks for walls or benchtops
£154.00 - £470.00
Product Code | Brand | Depth (English) | Product Type | Height (English) | Positions | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Depth (English) | Product Type | Height (English) | Positions | Price | Quantity & Availability | ||
11872801
|
Bel-Art™
F18933-0022 |
10 in. | Benchtop Rack (2-tier) | 15.4 in. | 76 |
£468.00
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||
15540102
|
Bel-Art™
F18933-0015 |
7 in. | Benchtop Rack (3-tier) | 22.4 in. | 57 |
£264.19
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
||
15530102
|
Bel-Art™
F18933-0012 |
7 in. | Benchtop Rack (2-tier) | 15.4 in. | 38 |
£271.00
Each |
In Stock
Log in to see stock. |
||
11862801
|
Bel-Art™
F18933-0021 |
10 in. | Benchtop Rack (1-tier) | 8.4 in. | 38 |
£222.00
Each |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||
11803001
|
Bel-Art™
F18933-0011 |
5 in. | Benchtop/Wall-mount Rack (1-tier) | 9.4 in. | 19 |
£191.00
Each |
Estimated Shipment: 27-11-2024
Log in to see stock. |
||
11302754
|
Bel-Art™
F18933-0013 |
5 in. | Wall-mount Rack (3-tier) | 23.4 in. | 57 |
£180.00
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
||
11866331
|
Bel-Art™
F18933-0014 |
5 in. | Wall-mount Rack (2-tier) | 16.4 in. | 38 |
£154.00
Each |
Estimated Shipment: 02-09-2024
Log in to see stock. |
||
11882801
|
Bel-Art™
F18933-0023 |
10 in. | Benchtop Rack (3-tier) | 22.4 in. | 114 |
£470.00
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
||
Description
Glassware and plasticware drying racks for walls or benchtops
- Dry and store delicate labware including beakers, flasks, bottles, test tubes and cylinders on these space-saving drying racks
- Single-sided benchtop models fit flat against a wall
- Double-sided benchtop models provide double the capacity (up to 114 pegs) with a slightly larger footprint
- Three sizes and lengths of V-channel pegs can be repositioned to hold a variety of labware
- Rugged ABS plastic body and pegs wipe clean and won't corrode
- Wall-mount models have mounting points that fit standard pegboards
- Pegs available separately to reconfigure rack for specific drying requirements
Specifications
ABS | |
37.5 cm | |
14.75 in. |