missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Labeled Wash Bottles
Click to view available options
Capacity:
16 oz., 500 mL
For Use With (Application):
Acetone
Isopropanol
Water
Description
Includes
Safety labeled wash bottles include Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemmical Abstract Service (CAS), protective equipment symbols and target organ information.
- Easy-to-read printed bottles
- Wide mouth 53mm (2 in.) bottle for easy refilling
- Translucent, LDPE solvent-resistant
- Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
- Toluene is red LDPE bottle
- Polypropylene closure with leakproof design
- Bottles can be purchased separately or in variety packs that include water, acetone, methanol, isopropanol, toluene and ethanol
Compliance
OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Specifications
Specifications
Capacity | 16 oz., 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Natural |
For Use With (Application) | Water |
Material | LDPE, Polyethylene |
Closure Color | Blue |
Autoclavable | Not Autoclavable |
Closure Material | Polypropylene |
Closure Size | 53 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction