missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Microscope Slide Tray

White polystyrene background provides good visual contrast for viewing dyes and stains

Brand:  Bel-Art™ H44581-0000

Product Code. 10126903

  • £134.00 / Pack of 6
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

White polystyrene background provides good visual contrast for viewing dyes and stains

  • Stores slides in a horizontal position for full visibility
  • Handy finger cut-out simplifies removal of slides
  • Each tray stores 20 standard 1 x 3 in. (25 x 75mm) slides
  • Can be used independently or conveniently stored in the Slide Tray Cabinet (Mfr. No. 445830000, sold separately)
  • Dimensions: 6.5 x 13.5 in. (17 x 34cm)

Specifications

Specifications

Slide Storage Tray
White
Microscope Slide
20
20
Polystyrene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Microscope Slide Tray > Stores 20 standard 1 x 3 in. (25 x 75mm) slides

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.