Learn More
Bel-Art™ SP Scienceware™ Mini Sampler Spoons
Avoid cross contamination by using a different spoon for each sample
Brand: Bel-Art™ F36721-0050
Code : NEW
Additional Details : Weight : 0.02340kg
Description
- These disposable mini measures are designed for retrieving small, specific volume samples of powders or similar materials
- Made from white high-impact polystyrene plastic
- All sterile products are lot stamped for accurate tracing
Specifications
Spoon | |
Rounded bowl and thin handle neck | |
Yes | |
White |
Polystyrene | |
0.5 mL | |
Sterile | |
Designed for retrieving small, specific volume samples of powders or similar materials |