missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Multiple Chromatography Column Holder

Bendable plastic loops gently support each column without scratching

Brand:  Bel-Art™ F18202-0000

168.15 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


Product Code. 10305313

  • £177.00 / Each
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket

Requires : Two support rods and stands. Order separately Cat. No. 22-260469.
This item is not returnable. View return policy

Description

Description

  • Securely holds as many as eight chromatography columns in or between its flexible loops at an adjustable height
  • The rigid plastic back is mounted on two polyethylene plastic support blocks which slide over support stand rods
  • Tighten the screws to mount the rack at the desired height
  • Holds columns up to 30mm (1.19 in.) in diameter

Specifications

Specifications

30 mm
311 mm
Chromatography Column Holder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Multiple Chromatography Column Holder > Column Holders

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.