missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ OdoClave™ Deodorant Autoclave Pads

Product Code. p-182263 Shop All SP Scienceware Products
Click to view available options
Scent:
Cherry
Clove
Lemon
Pine
Color:
Amber
Brown
Green
Red
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 11976072

Brand: Bel-Art™ H131980000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Neutralize autoclave odors with a pleasant fragrance

  • Heat-release deodorant works automatically at autoclave temperatures
  • Open pouch and place in bag prior to autoclaving
  • Three-year shelf life or longer if stored in a sealed container
  • 32mm (1.75 in.) square
  • Triple strength pads release three times the deodorant of standard pads while costing less than two standard pads

Order Info

Choose 1.25 in. (32mm) square regular-strength or 2.25 in. (5.7cm) square super-strength pad.

Specifications

Product Type Autoclave Capsule
Color-coded No
Form Liquid
Scent Clove
Sterility Non-sterile
Autoclavable Autoclavable
Color Brown
Format Pad
For Use With (Application) Autoclave
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.