Learn More
Bel-Art™ SP Scienceware™ Pipet Vacuum Aspirator Collection System
Performs vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes
Brand:  Bel-Art™ F19917-0155
79.57 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.
 View more versions of this product
Product Code. 15237513
  • £94.71 / Pack of 12
Estimated Shipment: 05-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Built-in pump quickly aspirates fluids from pipet tip, Pasteur pipet or serological pipet inserted into the collection tube
  • Maintains low noise level (76db at 1 meter) and minimal vibration
  • Excellent for fluid collection from tissue and/or cell culture, Petri dishes, plates, flasks and tubes
  • Chemical and corrosion resistant polypropylene collection bottle holds 3800mL (1 gallon) fluid wastes and has a safety release vent to prevent vacuum implosion
  • A replaceable hydrophobic filter seals itself if it becomes wet protecting the pump motor from fluid contamination
  • Lightweight and compact design will not crowd workspace in a hood or at the bench
Specifications
Specifications
Replacement Hydrophobic Filters
3800 mL