Learn More
Bel-Art™ PLUM™ Emergency Eyewash Station Refills
Refill kits for PLUM station
£30.70
Specifications
For Use With (Equipment) PLUM Eyewash Stations or Alone
View More Specs
Products 1
Product Code Brand Capacity (Metric) Includes Product Type Price Quantity & Availability  
Product Code Brand Capacity (Metric) Includes Product Type Price Quantity & Availability  
12719706
View Documents
Bel-Art™
F24880-0053
500 mL Three 500 mL bottles of saline solution, form-fitting eye cup, a dust cap and instructions Saline Eyewash Solution Refill This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Refill kits for PLUM station
  • Bottles contain a sterile 0.9% sodium chloride solution (saline) for rinsing dust and debris, or a 4.9% sterile phosphate buffer (pH neutralizing) solution to quickly neutralize acids and alkalis
  • Bottles have clear first aid instructions, twist-open design for quick access and a built-in eye cup with dust cap
  • Use to refill eye wash stations or take to job sites as part of first aid kits
  • Bottles require no maintenance and have a three-year shelf life from date of manufacture
Compliance
Meet all U.S. federal regulations. Comply with ANSI™ standards. NDC# and expiration date printed on every bottle.
Specifications
Specifications
PLUM Eyewash Stations or Alone
Videos
Safety and Handling
Safety and Handling
missing translation for 'shelfLife' : 3 Years