Learn More
Bel-Art™ SP Scienceware™ Poxygrid™ Baskets
Epoxy-coated steel wire construction holds up under severe laboratory conditions
Brand:  Bel-Art™ H16765-0000
 View more versions of this product
Product Code. 11773207
  • £129.00 / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Epoxy-coated steel wire construction holds up under severe laboratory conditions
  • Resist rust, corrosion, solvents, acids and alkalis
  • Convenient for transporting and batch-dipping glassware
  • Steam autoclavable at 121°C (250°F)
Specifications
Specifications
Epoxy-coated Steel
28 cm
15 cm
8.8 cm
11 x 6 x 3.5 in. (28 x 15 x 8.8cm)
11 in.
6 in.
3.5 in.