missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Bench-Top Biohazard Bag Holder

Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.

Brand:  Bel-Art™ F13193-0102

18.02 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.


 View more versions of this product

Product Code. 13472509

  • £19.50 / Each
Estimated Shipment: 07-07-2025
to see stock.
Explore more special offers
Add to basket

Includes: Plastic Cover Only
This item is not returnable. View return policy

Description

Description

Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.

  • Rectangular steel wire frame to hold one polyethylene bag of dimensions 8.5 x 11 in. (22 x 28cm)
  • Available with epoxy coat in orange color
  • Comes with snug fitting polypropylene plastic cover to hold bag securely in place
  • Reusable and allows cleaning by hand or in automated dish washers
  • Autoclavable
  • Also used with Clavies Autoclavable Bags H13182-0812

Order Info

For complete kit with bag holder, cover and biohazard bags, see Cat. No. 11-394-310.

Specifications

Specifications

Bag Holder Cover
Orange
Store, transport, or ship biohazardous wastes
Snug-Fitting Plastic Cover Only
5 in.
Autoclavable
Single-use
Biohazard bag holder
Polypropylene
12.7 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Bench-Top Biohazard Bag Holder > Cover for Bag Holder

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.