Learn More
Bel-Art™ Bench-Top Biohazard Bag Holder
Shop alle SP Scienceware produkterBeskrivelse
Indeholder
Cover, Bag Holder, and 100 Bench Top Biohazard Bags, made of high molecular weight, high-density (HMHD) Polyethylene and printed with Biohazard symbol and sterilization indicator patch that darkens when exposed to steam sterilization.
Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use.
- Rectangular steel wire frame to hold one polyethylene bag of dimensions 8.5 x 11 in. (22 x 28cm)
- Available with epoxy coat in orange color
- Comes with snug fitting polypropylene plastic cover to hold bag securely in place
- Reusable and allows cleaning by hand or in automated dish washers
- Autoclavable
- Also used with Clavies Autoclavable Bags H13182-0812
Overholdelse
When used with lid, provides compliance with 29 CFR 1910.1030(d)(2)(xiii) (A), which requires a cover on any container used to store, transport or ship biohazardous wastes.
Tekniske data
Tekniske data
| Autoclavable | Autoclavable |
| Color | Pink |
| Disposable | Single-use |
| For Use With (Application) | Perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use |
| For Use With (Equipment) | 01-828A, 01-828-031, 01-830A, 01-826-1, 01-827A, 01-829A, 01-815-1 22 x 28cm biohazard bag, Clavies™ autoclavable bags (01-815-6) |
| Includes | Complete Kit: Bag Holder, Cover and Bags (100pk) |
| Material | Polypropylene |
| Width (English) | 5 in. |
| Width (Metric) | 12.7 cm |
| Product Type | Biohazard Bag Holder Kit |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.